

Copper(I) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7758-89-6
:
Formula:ClCu
MDL Number:MFCD00010971
Merck Index:13,02690
Boiling Pt:1367 °C (1013 hPa)
Melting Pt:430 °C
Density:4.145 g/cm³ (25 °C)
EINECS:231-842-9
UN:2802
ADR:8,III
:M-Clarity™ quality level = MQ200
:99 g/mol
:GHS07
:GHS09
Storage Temperature:Ambient