Order Entry
Germany
ContactUsLinkComponent
Copper(I) chloride 97%, Reagent Grade
Irritant
Corrosive
Copper(I) chloride 97%, Reagent Grade
Certificates
Irritant
Corrosive
Copper(I) chloride 97%, Reagent Grade

Specification Test Results

  • Appearance of the substance:
    complying
  • Assay of Cu:
    61.9 - 66.4 %
  • Vapor Pressure:
    1.3 mmHg ( 546 °C)

Specification Test Results

Related Information