Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Copper (I) chloride
Copper(I) chloride ≥99% (metals basis)
Copper(I) chloride ≥99% (metals basis)
:
Thermo Fisher Scientific
Copper(I) chloride ≥99% (metals basis)
:
Thermo Fisher Scientific
:
7758-89-6
:
:
ClCu
:
MFCD00010971
:
14,2660
:
1490 °C
:
430 °C
:
4.14
:
Ambient
:
231-842-9
:
8,III
:
99 g/mol
:
2802
:
GHS07
:
GHS09
msds_ALFA041837
Thermo Scientific Acroseal 25 Years