

Copper(I) chloride ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-89-6
:
Formula:ClCu
MDL Number:MFCD00010971
Merck Index:14,2660
Boiling Pt:1490 °C
Melting Pt:430 °C
Density:4.14
Storage Temperature:Ambient
EINECS:231-842-9
ADR:8,III
:99 g/mol
UN:2802
:GHS07
:GHS09