


Copper(I) chloride 97%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-89-6
:
Formula:ClCu
MDL Number:MFCD00010971
Merck Index:13,02690
Boiling Pt:1367 °C (1013 hPa)
Melting Pt:430 °C
Density:4.14 g/cm³
Storage Temperature:Ambient
EINECS:231-842-9
ADR:8,III
:99 g/mol
UN:2802
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:Green-grey Powder or pearls
- Titration Complexometric:≥96.0 %
- Sulfate (SO4):≤500 ppm
- Iron (Fe):≤400 ppm