2,3-Dihydroxynaphthalene
Catalog # MOLEM82734472
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:160.17 g/mol
Boiling Pt:354 °C (1013 hPa)
Melting Pt:151…152 °C
Density:1,12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-156-7
CAS Number:92-44-4
Formula:C₁₀H₈O₂
MDL Number:MFCD00004073