



Ammonium cerium(IV) nitrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:16774-21-3
:
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121
:548.22 g/mol
Merck Index:13,00509
Melting Pt:107…108 °C
Density:2.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-827-6
ADR:5.1 (8),II
:M-Clarity™ quality level = MQ300
UN:3085
:GHS03
:GHS05
:GHS07
:GHS09
Specification Test Results
- Solubility:1410 g/l (20 °C)
- Melting Point:108 °C
- Molar Mass:548.22 g/mol
- Bulk Density:1200 kg/m3
- Density:2.49 g/cm3
- pH:1 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer