7-Methoxyindole
Catalog # MOLEM89429300
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:147.18 g/mol
Boiling Pt:298 °C (1013 hPa)
Density:1,126 g/cm³ (25 °C)
EINECS:221-690-1
:GHS07
Storage Temperature:Freezer
CAS Number:3189-22-8
Formula:C₉H₉NO
MDL Number:MFCD00047203
Boiling Pt:298 °C (1013 hPa)