Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methoxyindole
6-Methoxyindole
6-Methoxyindole
MOLE17682700-1G
:
Molekula
:
6-Methoxyindole
MOLE17682700-1G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
147.18 g/mol
:
221-689-6
:
GHS07
:
Freezer
:
3189-13-7
:
C₉H₉NO
:
MFCD00022780