6-Methoxyindole
Catalog # ABCRAB168523.0005
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:147.18 g/mol
EINECS:221-689-6
:GHS07
Storage Temperature:Freezer
CAS Number:3189-13-7
Formula:C₉H₉NO
MDL Number:MFCD00022780