Order Entry
Germany
ContactUsLinkComponent
 
Restriction enzymes, PfoI, Fermentas
Catalog # ER1751
CAS Number:  
undefined
Restriction enzymes, PfoI, Fermentas
Catalog # ER1751
CAS Number:  

Specifications

  • Concentration:
    10 U/µl
  • Pk:
    1 KIT
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    5' overhang
  • Description:
    PfoI
  • Enzyme name:
    PfoI
  • Units:
    200 units

Specifications

About this item

PfoI restriction enzyme recognises T^CCNGGA sites and cuts best at 37 °C in Tango buffer.

  • Stringent quality control and industry leading manufacturing process
  • Colour-coded five buffer system
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities

Its application includes molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP)and SNP.

PdoI is CpG methylation-sensitive, dam methylation-sensitive, dcm methylation-sensitive and also sensitive to heat inactivation.

Delivery: It includes universal Tango buffer for double-digestions.