Order Entry
Germany
Orders LinkContactUsLinkComponent
5-Iodoindole

5-Iodoindole

Glass bottle for solids, 5 g
Catalog # MOLE12190031-5G
Irritant
Marketsource
Supplier:   Molekula
5-Iodoindole
Catalog # MOLE12190031-5G
Supplier:  Molekula
Marketsource
CAS Number:  16066-91-4
5-Iodoindole

Glass bottle for solids, 5 g

CAS Number:  16066-91-4
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:16066-91-4
  • Formula:C₈H₆IN
  • MDL Number:MFCD00220065
  • Molecular Weight:243.05 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications