Indole-3-carboxylic acid
Catalog # MOLEM17920095
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:161.16 g/mol
EINECS:212-231-6
CAS Number:771-50-6
Formula:C₉H₇NO₂
MDL Number:MFCD00005624