Ninhydrin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:485-47-2
:
Formula:C₉H₆O₄
MDL Number:MFCD00003791
:178.14 g/mol
Merck Index:13,06582
Boiling Pt:351 °C (1013 hPa)
Melting Pt:250…258 °C
Density:0.862 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:207-618-1
:GHS07
Storage Temperature:Ambient