Ninhydrin ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:485-47-2
:
Formula:C₉H₆O₄
MDL Number:MFCD00003791
:178.14 g/mol
Merck Index:13,06582
Boiling Pt:351 °C (1013 hPa)
Melting Pt:250…258 °C
Density:0.862 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:207-618-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:20 g/l (20 °C)
- Melting Point:250 - 258 °C (decomposition)
- Molar Mass:178.15 g/mol
- Bulk Density:680 kg/m3
- pH:4.6 - 5.0 (10 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06762.0010_TDS_EN MERC1.06762.0100_TDS_EN ProductPage_106762 ProductPage_106762
Product specification
msds_106762