1,1'-Carbonyldiimidazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:530-62-1
:
Formula:C₇H₆N₄O
MDL Number:MFCD00005286
:162.15 g/mol
Merck Index:14,01819
Melting Pt:111…116 °C
Storage Temperature:Ambient
EINECS:208-488-9
UN:3263
ADR:8,II
:GHS05
:GHS07