Order Entry
Germany
ContactUsLinkComponent
1,1'-Carbonyldiimidazole for synthesis, Sigma-Aldrich®
1,1'-Carbonyldiimidazole for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1,1'-Carbonyldiimidazole for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    116 - 120 °C
  • Molar Mass:
    162.15 g/mol
  • pH:
    7.7 (50 g/l, H2O, 20 °C)
  • Hill Formula:
    C7H6N4O

Specification Test Results

Related Information