1,1'-Carbonyldiimidazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:530-62-1
:
Formula:C₇H₆N₄O
MDL Number:MFCD00005286
:162.15 g/mol
Merck Index:14,01819
Melting Pt:118…120 °C
Storage Temperature:Refrigerator
EINECS:208-488-9
UN:3259
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Assay:≥96.0 %
- Water:≤0.5 %
- GC:≥97.5 %