Order Entry
Germany
ContactUsLinkComponent
1,2-Dimethylimidazole for synthesis, Sigma-Aldrich®
Corrosive
Irritant
1,2-Dimethylimidazole for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
1,2-Dimethylimidazole for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    30 - 40 °C
  • Molar Mass:
    96.13 g/mol
  • Boiling Point:
    205 °C (1013 hPa)
  • Vapor Pressure:
    1 hPa (20 °C)
  • Flash Point:
    93 °C
  • Density:
    1.08 g/cm3 (20 °C)
  • pH:
    12.6 (500 g/l, H2O, 20 °C)
  • Ignition Point:
    480 °C

Specification Test Results

Related Information