
Xenon difluoride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13709-36-9
:
Formula:F₂Xe
MDL Number:MFCD00040538
:169.29 g/mol
Merck Index:14,10072
Boiling Pt:114 °C (1013 hPa)
Density:4.32 g/cm³ (20 °C)
EINECS:237-251-2
ADR:5.1,II
:M-Clarity™ quality level = MQ100
UN:3087
:GHS05
Storage Temperature:Ambient