Iodine(V) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12029-98-0
:
Formula:I₂O₅
MDL Number:MFCD00011356
:333.81 g/mol
Merck Index:12,05040
Density:4.79 g/cm³ (20 °C)
EINECS:234-740-2
UN:1479
ADR:5.1,III
:M-Clarity™ quality level = MQ100
:GHS03
:GHS07