Iodine(V) oxide analytical reagent, Supelco®
1.00358.0100
: Merck
:
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
- Solubility:(20 °C) soluble
- Molar Mass:333.81 g/mol
- Bulk Density:2000 kg/m3
- Density:4.98 g/cm3 (20 °C)
- pH:1.1 (100 g/l, H2O, 25 °C)
:
:333.81 g/mol
:12,05040
:4.79 g/cm³ (20 °C)
:5.1,III
:M-Clarity™ quality level = MQ100
:234-740-2
:1479
:GHS03
:GHS07
:12029-98-0
:I₂O₅
:MFCD00011356