Order Entry
Germany
ContactUsLinkComponent
Iodine trichloride (Wijs'reagent) for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Iodine trichloride (Wijs'reagent) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Iodine trichloride (Wijs'reagent) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    60 °C (decomposition)
  • Molar Mass:
    233.26 g/mol
  • Vapor Pressure:
    1.3 hPa (64 °C)
  • Density:
    3.12 g/cm3 (20 °C)
  • Hill Formula:
    Cl3I
  • Chemical Formula:
    ICl3

Specification Test Results

Related Information