Dimethyl sulphoxide
Catalog # UPST20-139
Supplier: UPSTATE TECHNOLOGY
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:4 mL
- Seq:2195
Specifications
About this item
:
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089