Bottle-top dispensers, Dispensette® S Organic, DAkkS
Supplier: Brand
Certificates
About this item
Analogue-adjustable or Digital Easy Calibration.
- With or without recirculation valve
- Autoclavable at 121 °C
- Model of volume adjustment available: Analog (Analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
- Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy Calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
- DE-M marking
- Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated
Dispensette® S Organic is ideal for dispensing organic solvents: such as chlorinated and fluorinated hydrocarbons like trichlorotrifluoroethane and dichloromethane, or acids like concentrated HCl and HNO3 (except for HF), as well as for trifluoroacetic acid (TFA), tetrahydrofurane (THF), and peroxides. For dispensing hydrofluoric acid (HF), we recommend the use of the Dispensette® S Trace Analysis bottle-top dispenser with platinum-iridium valve spring. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, tantalum, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: Yellow.
Ordering information: Dispensette® S Organic supplied with DAkkS certificate, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and adapters of PP.