
Amidosulphonic acid, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:5329-14-6
:
Formula:H₃NO₃S
MDL Number:MFCD00011603
:97.09 g/mol
Merck Index:13,09006
Boiling Pt:247 °C (1013 hPa)
Melting Pt:205 °C
Density:2.15 g/cm³ (20 °C)
UN:2967
ADR:8,III
:M-Clarity™ quality level = MQ100
EINECS:226-218-8
:GHS07
Storage Temperature:Ambient