
Amidosulphonic acid 99%
:5329-14-6
:
:H₃NO₃S
:MFCD00011603
:97.09 g/mol
:13,09006
:247 °C (1013 hPa)
:200…208 °C
:2,15 g/cm³ (20 °C)
:Ambient
:226-218-8
:2967
:8,III
:GHS07
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:200 to 208 °C
- Titration with NaOH:≥98.5 %