Order Entry
Germany
ContactUsLinkComponent
Ammonium hexafluorosilicate, Sigma-Aldrich®
Toxic
Ammonium hexafluorosilicate, Sigma-Aldrich®
Catalog # SIAL204331-50G
Supplier:  Merck
CAS Number:  
Toxic
Ammonium hexafluorosilicate, Sigma-Aldrich®
Catalog # SIAL204331-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
Merck Index:13,00529
Density:2.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-968-3
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
 :178.15 g/mol
UN:2854
 :GHS06
CAS Number:16919-19-0
Formula:H₈F₆N₂Si
MDL Number:MFCD00010887