Diammonium hydrogen orthophosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7783-28-0
:
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891
:132.06 g/mol
Merck Index:13,00546
Melting Pt:185 °C
Density:1.619 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-987-8
REACH:01-2119490974-22