Order Entry
Germany
ContactUsLinkComponent
Diammonium hydrogen orthophosphate ≥98%
Diammonium hydrogen orthophosphate ≥98%
Catalog # A17416.36
CAS Number:  
Diammonium hydrogen orthophosphate ≥98%
Catalog # A17416.36
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :132.06 g/mol
Merck Index:14,542
Melting Pt:155 °C (dec.)
Density:1.619
EINECS:231-987-8
REACH:01-2119490974-22
 :GHS07
Storage Temperature:Ambient
CAS Number:7783-28-0
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891