Diammonium hydrogen orthophosphate ≥98.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7783-28-0
:
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891
:132.06 g/mol
Merck Index:14,542
Melting Pt:155 °C (dec.)
Density:1.619
EINECS:231-987-8
REACH:01-2119490974-22
:GHS07
Storage Temperature:Ambient