Order Entry
Germany
ContactUsLinkComponent
Ammonium hydrogen carbonate ≥97%, GPR RECTAPUR®
Irritant
Ammonium hydrogen carbonate ≥97%, GPR RECTAPUR®
Catalog # 21218.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonium hydrogen carbonate ≥97%, GPR RECTAPUR®
Catalog # 21218.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.05 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :79.06 g/mol
Merck Index:13,00497
Melting Pt:35 °C
Density:1.57 g/cm³ (20 °C)
EINECS:213-911-5
 :GHS07
CAS Number:1066-33-7
Formula:NH₄HCO₃
MDL Number:MFCD00012138