
Ammonium hydrogen carbonate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1066-33-7
:
Formula:NH₄HCO₃
MDL Number:MFCD00012138
:79.06 g/mol
Merck Index:14,497
Melting Pt:∼ 60 °C (dec.)
Density:1.586
EINECS:213-911-5
:GHS07
Storage Temperature:Ambient