Order Entry
Germany
ContactUsLinkComponent
 
 
Ammonium hydrogen carbonate 98%, pure
 
undefined
 
Ammonium hydrogen carbonate 98%, pure
 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration with HCl:
    ≥97.5 %
  • Anticaking agent:
    ≤0.55 % (MgCO3)