

Ammonium carbonate, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1.500 g/cm³ (20 °C)
EINECS:208-058-0
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Assay (NH3):30.0 - 34.0 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 50 ppm
- Insolubility in water:Max. 100 ppm
- Cl (Chloride):Max. 5 ppm
- NO3 (Nitrate):Max. 10 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- As (Arsenic):Max. 3 ppm
- Fe (Iron):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
Packaging Information
21217_AMMONIUM CARBONATE ANALAR NORMAPUR_BSE_REV0
BSE/TSE Certificates
spec_VWRC21217.260 spec_VWRC21217.260 spec_VWRC21217.295 spec_VWRC21217.295
Product specification
VWR® for chromatography columns, consumables and reagents