Ammonium carbonate, EMPROVE® ESSENTIAL Ph. Franç., NF, E503, SAFC®
Catalog # 1.01136.9051
Supplier: Merck
CAS Number:
Specifications
- Pack type:Cardboard carton
- Pk:50 kg
- Seq:4418
Specifications
About this item
:
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1.5 g/cm³ (20 °C)
EINECS:233-786-0
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:506-87-6
:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890