Order Entry
Germany
ContactUsLinkComponent
Triphenylphosphine ≥99%
Triphenylphosphine ≥99%
Catalog # ABCRS15-6700.0100
Supplier:  abcr
CAS Number:  
Triphenylphosphine ≥99%
Catalog # ABCRS15-6700.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :262.29 g/mol
Merck Index:12,09873
Boiling Pt:360 °C
Melting Pt:79 °C
Density:1.194 g/cm³ (20 °C)
Flash Pt:359 °F
EINECS:210-036-0
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:603-35-0
Formula:C₁₈H₁₅P
MDL Number:MFCD00003043