Order Entry
Germany
ContactUsLinkComponent
Triphenylphosphine
Triphenylphosphine
Catalog # EHERC17893300
Supplier:  EHRENSTORFER
CAS Number:  
Triphenylphosphine
Catalog # EHERC17893300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :262.29 g/mol
Merck Index:12,09873
Boiling Pt:377 °C (1013 hPa)
Melting Pt:79 °C
Density:1.194 g/cm³ (20 °C)
Flash Pt:180 °C
EINECS:210-036-0
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:603-35-0
Formula:C₁₈H₁₅P
MDL Number:MFCD00003043