Triphenylphosphine 99%, powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:603-35-0
:
Formula:C₁₈H₁₅P
MDL Number:MFCD00003043
:262.29 g/mol
Merck Index:14,9743
Boiling Pt:360 °C
Melting Pt:79…81 °C
Density:1.200
Flash Pt:181 °C (358 °F)
Storage Temperature:Ambient
EINECS:210-036-0
:GHS08
:GHS07