
Acetylcholine iodide for synthesis, Sigma-Aldrich®
Catalog # 8.41870.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Melting Point:162 - 165 °C
- Molar Mass:273.11 g/mol
Specification Test Results
About this item
:
:273.11 g/mol
Melting Pt:163…165 °C
Density:1.67 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:218-862-3
:GHS07
Storage Temperature:Freezer
CAS Number:2260-50-6
Formula:H₃CC(O)O(CH₂)₂N(CH₃)₃I
MDL Number:MFCD00011815