Order Entry
Germany
ContactUsLinkComponent
Choline chloride
Choline chloride
Catalog # MOLEM16444475
Supplier:  Molekula
CAS Number:  
undefined
Choline chloride
Catalog # MOLEM16444475
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :139.62 g/mol
Merck Index:13,02226
Melting Pt:244…247 °C
Density:∼1.205 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-655-4
CAS Number:67-48-1
Formula:HOH₂CCH₂N(CH₃)₃Cl
MDL Number:MFCD00011721