Order Entry
Germany
ContactUsLinkComponent
 
Choline chloride ≥98.0%, white crystalline powder
 
undefined
Choline chloride ≥98.0%, white crystalline powder

 

  • Identity Test Passes:
    Passes
  • Assay:
    ≥98,0% (anhydrous basis)
  • Heavy Metals:
    ≤10 ppm (as Pb)
  • pH:
    4 - 8
  • Residue on Ignition:
    ≤0,1%
  • Water Content:
    ≤0,5%
  • Solubility (10% aq soln) :
    Clear and Colorless
  • 1,4-Dioxane:
    ≤10 ppm