Choline chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-48-1
:
Formula:HOH₂CCH₂N(CH₃)₃Cl
MDL Number:MFCD00011721
:139.62 g/mol
Merck Index:13,02226
Melting Pt:298…304 °C (dec.)
Density:~ 1,205 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-655-4
Specification Test Results
- Appearance:White Adhering crystals
- Infrared spectrum:Conforms
- Titration Argentometric:98.5 to 101.5 % (on dry substance)
- Water:≤2 %