
Tetrabutylammoniumchlorid zur Synthese, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1112-67-0
:
Formula:C₁₆H₃₆ClN
MDL Number:MFCD00011635
:277.92 g/mol
Melting Pt:83…86 °C
Density:1,05 g/cm³ (20 °C)
:M-Clarity™ Qualitätsstufe = MQ200
Flash Pt:110 °C
EINECS:214-195-7
:GHS07
Specification Test Results
- Melting Point:41 - 44 °C
- Molar Mass:277.92 g/mol
- Bulk Density:700 kg/m3
- Flash Point:110 °C
- pH:5 - 8 (100 g/l, H2O, 20 °C)
- Hill Formula:C16H36ClN
Specification Test Results
Frequently Bought Together