Order Entry
Germany
ContactUsLinkComponent
Tetraethylammonium bromide
Tetraethylammonium bromide
Catalog # MOLEM26739038
Supplier:  Molekula
CAS Number:  
Tetraethylammonium bromide
Catalog # MOLEM26739038
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :210.16 g/mol
Merck Index:13,09274
Melting Pt:> 285 °C
Density:1.43 g/cm³ (20 °C)
EINECS:200-769-4
CAS Number:71-91-0
Formula:C₈H₂₀BrN
MDL Number:MFCD00011825