Tetraethylammonium bromide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:71-91-0
:
Formula:C₈H₂₀BrN
MDL Number:MFCD00011825
:210.16 g/mol
Merck Index:13,09274
Melting Pt:> 285 °C
Density:1.43 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:200-769-4
Specification Test Results
- Solubility:2795 g/l (25 °C)
- Melting Point:285 °C
- Molar Mass:210.16 g/mol
- Density:1.397 g/cm3 (20 °C)
- pH:6.5 (100 g/l, H2O, 20 °C)