
Tetraethylammonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-91-0
:
Formula:C₈H₂₀BrN
MDL Number:MFCD00011825
:210.16 g/mol
Merck Index:14,9199
Melting Pt:∼ 285 °C (dec.)
Density:1.397
EINECS:200-769-4
:GHS07
Storage Temperature:Ambient