Tetraethylammonium bromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-91-0
:
Formula:C₈H₂₀BrN
MDL Number:MFCD00011825
:210.16 g/mol
Merck Index:13,09274
Melting Pt:285 °C (dec.)
Density:1,43 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-769-4
Specification Test Results
- Appearance:White to cream Crystalline powder or crystals or solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Loss on drying:≤2 %