Cetrimonium bromide
Supplier: Molekula
Certificates
About this item
CAS Number:57-09-0
:
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772
:364.45 g/mol
Merck Index:13,02034
Melting Pt:237…243 °C
Flash Pt:244 °C (closed cup)
EINECS:200-311-3
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS05
:GHS09
Storage Temperature:Ambient