Order Entry
Germany
ContactUsLinkComponent
 
 
Triisobutyl phosphate for synthesis, Sigma-Aldrich®
 :  Merck
 
 
Triisobutyl phosphate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    0.26 g/l (20 °C)
  • Molar Mass:
    266.32 g/mol
  • Boiling Point:
    272 °C (1013 hPa) decomposes
  • Vapor Pressure:
    2 hPa (103 °C)
  • Flash Point:
    133 °C DIN 51758
  • Density:
    0.965 g/cm3 (20 °C)
  • Ignition Point:
    430 °C DIN 51794